Learn More
Honeywell™ Safety Uvex™ Turboshield™ Visors
Replacement visor for use with hard hat adapter or headgear.
Supplier:  Honeywell™ S9575
 View more versions of this product
Catalog No. 19-830-115
Description
Description
  • Replacement visor
  • Clear
  • PC visor
  • Hardcoat/anti-fog
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.