Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Smitty™ Textured Nitrile Coated Seamless Knit Gloves

Catalog No. 19025828 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-025828 Medium
19-025829 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-025828 Supplier Honeywell™ Supplier No. 81/1162M
  • $972.00
    $486.65 / Case of 144

    Save $485.35

    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless knit construction minimizes irritation for improved user comfort, while the nitrile coating provides abrasion resistance and enhanced grip capabilities.

  • Seamless knit construction
  • Minimizes rubbing and chafing for increased user comfort
  • Nitrile coated palm provides abrasion resistance, and enhances grip capabilites
  • Knit wrist helps prevent dirt and debris from entering the glove

Specifications

Certifications/Compliance FDA 21 CFR 177.2600
Construction Seamless Knit
Coating Material Nitrile
Color Natural, Red
Exterior Finish Partially Textured
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Medium
For Use With (Application) Construction, Manufacturing, Municipal Services, and Transportation
Gender Unisex
Glove Cuff Style Knit
Grip Design Textured
Series 81/1162
Product Line Smitty
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.