Learn More
Description
Specifications
Specifications
| Antigen | COX IV |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Formulation | Buffer: PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3. |
| Gene Accession No. | P13073 |
| Host Species | Rabbit |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
