Learn More
Honeywell™ Whiting & Davis Metal Mesh Gloves
Description
Made of stainless-steel ring mesh; Highest level of cut protection; Reversible glove with rubber band straps for a snug fit.
Entire glove is made of a mesh of high tensile-strength, corrosion-resistant stainless-steel rings, each individually welded for maximum strength and remarkable flexibility. Provides the highest level of protection against cuts and lacerations in cutting operations. Ergonomic design offers optimal comfort and fit; fingers are tapered and glove body is contoured to the shape of the hand. Self-adjusting elastic band grips wrist snugly, holding glove in place. Reversibility maximizes use and reduces inventory costs. Stainless-steel rings resist process oils and fats, and are easy to clean.
Specifications
Specifications
| Material | Stainless Steel Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Medium |
| For Use With (Application) | Meat, poultry, fish processing, food service, knife and blade sharpening/cleaning, fishing, garment manufacturing, veterinary services, animal control, paper and pulp industry |
| Glove Cuff Style | Elasticated |