Learn More
Honeywell™ WorkEasy™ 300 Dyneema™ Cut-Resistant Gloves, Polyurethane Coated
Description
- High level of cut protection for hazardous applications
- Lightweight fabric blend provides the same level of protection, without the bulk to prevent hand fatigue
- Polyurethane coating provides abrasion resistance and enhanced grip
- Launderable for extended glove life
Specifications
Specifications
| Coating Material | Polyurethane |
| Color | Gray |
| Cut Resistance Rating | ANSI A3 |
| Exterior Finish | No Texture |
| Hand | Specific |
| Material | Dyneema Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Small |
| For Use With (Application) | Small parts assembly, fabrication, inspection, warehousing |
| Glove Cuff Color | White |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.