Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ WorkEasy™ 300 Dyneema™ Cut-Resistant Gloves, Polyurethane Coated

Catalog No. 19175488
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-175-488 X-Large
19-175-485 Small
19-175-486 Medium
19-175-487 Large
19-175-489 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-175-488 Supplier Honeywell™ Supplier No. WE300XL
This item is not returnable. View return policy

Dyneema™ fiber blend provides a high level of protection against cuts and abrasions, while still maintaining a bare-handed feel.

  • High level of cut protection for hazardous applications
  • Lightweight fabric blend provides the same level of protection, without the bulk to prevent hand fatigue
  • Polyurethane coating provides abrasion resistance and enhanced grip
  • Launderable for extended glove life
TRUSTED_SUSTAINABILITY

Specifications

Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A3
Exterior Finish No Texture
Hand Specific
Material Dyneema Fiber
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Small parts assembly, fabrication, inspection, warehousing
Glove Cuff Color White
Glove Cuff Style Knit
Model WE300-XL
Series WorkEasy 300
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.