Learn More
Enzo Life Sciences Honokiol (25mg). CAS: 35354-74-6
SDP
Supplier:  Enzo Life Sciences ALX350350M025
Encompass_Preferred
Potent and highly tolerable antitumor and antiangiogenic compound. Anxiolytic, anti-thrombotic and antibacterial. Shows central depressant effects in vivo. Inhibitor of nitric oxide (NO) and TNF-α production in LPS-activated macrophages by the suppression of inducible nitric oxide synthase (iNOS; NOS II) expression (IC50=6.4µM). Isomeric with magnolol (ALX-350-352). Inhibits NF-κB activation. Potent scavenger of superoxide and peroxyl radicals. Alternative name: 5,3'-Diallyl-2,4'-biphenyldiol, 5,3'-Diallyl-2,4'-dihydroxydiphenyl. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8855
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.