Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Honokiol, Anti-Inflammatory Chemotherapeutic Agent, Solid, 10 mg
Description
- Purity: >98%
- Molecular Weight: 266.34 g/mol
- Form: Solid
- CAS Number: 35354-74-6
- Key Properties: Potent scavenger of reactive oxygen species with antiangiogenic, antitumor and anti-inflammatory properties
- Mechanism of Action: Inhibition of ras signalling and activation of the mitochondrial transistion pore via cyclophilin D induction; inhibits NF-κB signaling, upstream of VEGF
- Characteristics: Blood-brain barrier permeable
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Buffer | 100% Honokiol |
| Formulation | 100% Honokiol |
| Quantity | 10 mg |
| For Use With (Application) | Anti-inflammatory, anti-tumor, anti-angiogenic research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?