Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Honokiol, Anti-inflammatory Chemotherapeutic Agent, Solid, 25 mg

Catalog No. 502707391
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7391 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7391 Supplier Abcam Supplier No. AB12064725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent reactive oxygen species scavenger with antiangiogenic, antitumor, and anti-inflammatory properties.

  • Purity: >98%
  • Molecular Weight: 266.34 g/mol
  • Form: Solid
  • Properties: Potent scavenger of reactive oxygen species; antiangiogenic, antitumor, and anti-inflammatory properties
  • Mechanisms of Action: Inhibition of Ras signaling, activation of the mitochondrial transition pore via cyclophilin D induction, and inhibition of NF-κB signaling (upstream of VEGF)
  • Other Attributes: Blood-brain barrier permeable
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Reactive Oxygen Species, Ras, Cyclophilin D, NF-κB, VEGF
Product Type Antioxidant
Buffer 100% Honokiol
Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Formulation 100% Honokiol
Quantity 25 mg
For Use With (Application) Cancer research, anti-inflammatory research, antiangiogenic research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.