Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Honokiol dichloroacetate | 1620160-42-0 | 99.1% | 488.19 g/mol | C22H18Cl4O4 | 10MG
SDP

Supplier:  Medchemexpress LLC HY12429210MG

Encompass_Preferred

Honokiol DCA (honokiol dichloroacetate) is a dichloroacetate analog of honokiol supplied for research use. It has been reported to inhibit growth of human prostate cancer cells in vitro and to suppress androgen receptor (AR) protein levels, and is commonly used in cellular and mechanistic studies.

  • Dichloroacetate analog of honokiol.
  • Reported to inhibit prostate cancer cell growth in vitro.
  • Suppresses androgen receptor protein levels.
  • Available as mg-scale solids and as 10 mM solution in DMSO.
  • High purity (about 99.1%).
  • Molecular weight 488.19 g/mol; formula C22H18Cl4O4.
  • For research use only; not for clinical use.

Catalog No. 50-002-37173


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.