Learn More
Description
- This face shield bag comfortably stores and protects Enespro hoods
- Drawstring Closure
- Care and maintenance printed on the inside bag
- Approx. 24 x 14.5 x 19 in.
Specifications
Specifications
| Width (English) | 14.5 in. |
| Size | Standard |
| Product Type | Hood Bag |
| Material | Cotton flannel |
| Color | Beige |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
