Learn More
Grainger Hook and Loop 1x2 In White PK400
Supplier:  Grainger 16U735
Encompass_Preferred
The HALCO USA Hook and Loop 825PF3 is a hook and loop fastener that is made of nylon and has an adhesive backing. It has a width of 1 inch and a length of 25 feet. The fastener has hooks on one side and loops on the other for attaching two surfaces together. It can withstand temperatures up to 200°F. The adhesive backing allows it to adhere to smooth surfaces. This industrial strength hook and loop fastener is designed for hanging tools, securing wires and more. It meets military specification MIL-F-21840.
Catalog No. 50-276-4915
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.