Learn More
Grainger Hook and Loop 1x6 In Black PK50
Supplier:  Grainger 16U742
Encompass_Preferred
The VELCRO Brand Hook and Loop Fastener Roll is a 2" wide x 15' long roll of black nylon hook and loop fastener. It features a hook side that adheres to loop material and a loop side that adheres to hook material, allowing for adjustable fastening and connection. This industrial strength fastener can withstand repeated use and washing. It bonds instantly, offers high shear and peel strength, and resists fluids, abrasion, and chemicals. Common uses include clothing, shoes, bags, automotive, crafts, and industrial applications. This versatile fastener provides easy fastening and unfastening for temporary or semi-permanent bonds.
Catalog No. 50-276-6106