Learn More
Grainger Hook and Loop 1x6 In White PK150
Supplier:  Grainger 16U747
Encompass_Preferred
The Scotch-Brite Hook and Loop Sanding Disc, 2FXD4, is a 6 inch diameter sanding disc with a hook and loop backing that attaches to sanders and buffers. It features an open web disc design and is made of high quality aluminum oxide mineral for aggressive sanding and finishing on metal, wood, drywall, and paint. This disc has a very fine grit of 600 grit. It can be used for light sanding, smoothing, and final finishing. The disc is long lasting and washable for reuse. It works on electric and pneumatic sanders. This Scotch-Brite sanding disc provides high performance sanding while reducing scratches.
Catalog No. 50-276-4909