Learn More
Grainger Hook and Loop 1x6 In White PK75
Supplier:  Grainger 16U746
Encompass_Preferred
The VELCRO Brand Hook and Loop Fastener Roll is a 2" wide x 15' long roll of industrial strength hook and loop fastener tape. It features a polyester hook side that adheres to the woven nylon loop side. This VELCRO brand fastener is designed for general purpose use and bonds on contact, allowing easy fastening and unfastening. It can withstand washings and exposure to sunlight, chemicals, and solvents. This hook and loop fastener roll can be cut to desired lengths.
Catalog No. 50-276-5389