Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Hook Collar Clamp
SDPSCI_ED

Catalog No. S90317A Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Equipment):
1/2 in. diameter rods
3/4 in. diameter rods
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S90317A 1/2 in. diameter rods
S90317B 3/4 in. diameter rods
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S90317A Supplier United Scientific™ Supplier No. HKCL01
Only null left
Add to Cart
Add to Cart

Hook Collar Clamp for suspending pulleys, balances, etc. from support rods or cross members.

  • Plated steel collar clamps
  • Thumbscrews and hooks for suspending pulleys, balances, etc. from support rods or crossbars.
  • For 1/2 in. to 3/4 in. diameter rods

Specifications

Type Hose and Tubing Clamp
Material Steel
For Use With (Equipment) 1/2 in. diameter rods
For Use With (Application) For suspending pulleys, balances etc. from support rods or cross members 0.5 in.
Description Hook Collar Clamp, for 1/2 in. diameter rods
Color Silver
Jaw Type Fixed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.