Learn More
Invitrogen™ HOOK3 Polyclonal Antibody

Description
Immunogen sequence: IALLQSLLDD ANLRKNELET ENRLVNQRLL EVQSQVEELQ KSLQDQGSKA EDSVLLKKKL EEHLEKLHEA NNELQKKRAI IEDLEPRFNN SSLKIEELQE ALRKKEEEMK QMEERYKKYL EKAKSVIRTL DPKQNQGAAP EIQALKNQLQ ERDRLFHSLE KEYEKTKSQR EMEEKYIVSA WYNMGMTLHK KAAEDRLAST GSGQSFLARQ RQATSSRRSY PGHVQPATAR.
Specifications
Specifications
| Antigen | HOOK3 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.68 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | HOOK3 |
| Gene Accession No. | Q86VS8, Q8BUK6 |
| Gene Alias | 5830454D03Rik; AI317159; E330005F07Rik; hHK3; h-hook3; HK3; hook homolog 3; hook homolog 3 (Drosophila); hook microtubule-tethering protein 3; Hook3; mHK3; protein Hook homolog 3 |
| Gene Symbols | HOOK3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.