Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Hooke's Law Apparatus
SDPSCI_ED

Catalog No. S139500 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139500 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139500 Supplier United Scientific™ Supplier No. HKLW12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Classic demonstration that is large enough for the entire class to see.

Quantitatively show the principles related to Hooke's Law with this complete apparatus.

  • Includes a 14 in. metal rod that mounts on a 4-1/2 in. diameter base
  • Scale in centimeters and inches can be easily adjusted along the rod
  • The plated steel weight holder and spring are attached to an adjustable collar with crossbar
  • Requires slotted weights (not included – United Scientific WHST08 set recommended)
  • Includes activity guide

Specifications

Grade 5 to 12
Format Kit
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.