Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Hooke's Law Spring Set
SDPSCI_ED

Catalog No. S27845 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Spring Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S27845 Spring Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27845 Supplier United Scientific™ Supplier No. SPGRS5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Set of 5 springs with varying spring constants

  • Forces required for a 1 cm extension are 0.25, 0.5, 1, 1.5, and 2.5 Newtons
  • Springs each feature a hook to support a weight hanger and a red arrow to indicate position
  • Pre-stress removed for accurate results
  • Activity guide included

Specifications

Product Type Spring Set
For Use With (Application) Hooke's Law
Includes Springs (5), Activity Guide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.