Learn More
United Scientific™ Hooked Aluminum Weight Set of 6
SDPSCI_ED
Catalog No. S139522 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
Set of 6
Catalog No. Quantity
S139522 Set of 6
1 options
This item is not returnable. View return policy
Catalog No. S139522 Supplier United Scientific™ Supplier No. WSAL06
Add to Cart
Add to Cart
This item is not returnable. View return policy
High quality and economically priced weight set.
  • Set of six hooked aluminum masses with a plastic storage base
  • Includes one each of 10 g, 20 g, 30 g, 40 g, 50 g, and 100 g masses
Specifications
Product Type Weight Set
Material Aluminum
Weight (Metric) 10, 20, 30, 40, 50, 100 g
Quantity Set of 6
Warning: Cancer and reproductive harm
Prop 65