Learn More
Description
- Recommened for use in the fields of food and protected environments
- Porous material allows for better breathability
- PVC sole is completely waterproof, and ultra resistant to high pressure cleaning
- Can be used indoors or outdoors
- Fully covers the entire boot
Specifications
Specifications
| Size | Universal |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Closure Type | Elastic |
| Thickness | 55 g/m2 |
| For Use With (Application) | Any Protected Environment |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.