Learn More
Description
- Lightweight polypropylene cap improves user's comfort
- Ability to use a color code for tracking
- Adapts to different head sizes
Specifications
Specifications
| Color | White |
| Disposable | Single-use |
| Packaging Type | Polybag |
| Construction | Sewn and Elasticated |
| For Use With (Application) | HACCP Processes and Various Industries |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.