Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medicom™ Hopen™ Classic Bouffant Cap
Click to view available options
Color:
White
Description
- Lightweight polypropylene cap improves user's comfort
- Ability to use a color code for tracking
- Adapts to different head sizes
Specifications
Specifications
Disposable | Single-use |
Construction | Sewn and Elasticated |
For Use With (Application) | HACCP Processes and Various Industries |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction