Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Classic Bouffant Cap

Catalog No. 19910699 Shop All Medicom Products
Change view
Click to view available options
Color:
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-910-699 -
19-910-698 -
19-910-697 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-910-699 Supplier Medicom™ Supplier No. 70.003
This item is not returnable. View return policy

Medicom™ Hopen™ Classic Bouffant Cap available in three distinct colors

  • Lightweight polypropylene cap improves user's comfort
  • Ability to use a color code for tracking
  • Adapts to different head sizes

Specifications

Disposable Single-use
Construction Sewn and Elasticated
For Use With (Application) HACCP Processes and Various Industries
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.