Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Collared Hygiene Gown

Catalog No. 19910630 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
White
Closure Type:
Press Studs
Zipper
Size:
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type Size
19-910-630 Blue Press Studs 2X-Large
19-910-628 Blue Press Studs Large
19-910-629 Blue Press Studs X-Large
19-910-625 White Zipper Large
19-910-626 White Zipper X-Large
19-910-627 White Zipper 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-910-630 Supplier Medicom™ Supplier No. 40.409/05
This item is not returnable. View return policy

Medicom™ Hopen™ Polypropylene Coveralls available in collared, or hooded styles

  • Pleasant, and light PP SMS fabric is breathable and solid
  • SMS fabric makes it possible to better limit the particulate release of fibers
  • Category 1 Personal Protective Equipment
  • Suitable for protected environments
  • Individually packaged for better hygiene and controlled distribution
  • Elastic thumb holes contain latex, a natural material that contains allergenic proteins
  • Dark blue gown has anti-static properties giving it the ability to dissipate electrostatic charges
    • Not intended for use in a flammable, oxygen-enriched atmosphere, or an ATEX area

Specifications

For Use With (Application) Category 1 Personal Protective Equipment
Construction Made in China
Certifications/Compliance (Europe) EN 1149-5
Size 2X-Large
Product Type General Purpose Gown
Packaging Type Individual, Bag
Gender Unisex
No. of Pockets None
Material SMS
Color Blue
Closure Type Press Studs
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Latex Free No
Seam Type Sewn
Certifications/Compliance ISO 9001
Weight (Metric) 50 gsm
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.