Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Double Elastic Bouffant cap

Catalog No. 19910689 Shop All Medicom Products
Change view
Click to view available options
Color:
Pink
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-910-689 -
19-910-690 Pink
19-910-688 White
19-910-687 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-910-689 Supplier Medicom™ Supplier No. 70.112

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Medicom™ Hopen™ Double Elastic Bouffant cap available in four distinct colors

  • Lightweight, polypropylene bouffant cap provides additional comfort the user
  • Full head coverage ensures excellent retention for the user
  • Ability to use a color code for tracking
  • Can be used to control the environment for HACCP processes and various industries

Specifications

Disposable Single-use
For Use With (Equipment) Operating Room PPE
Width (Metric) 50 cm
Construction Made in China
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.