Learn More
Medicom™ Hopen™ HOP'CLEAN Hygiene Coveralls
Catalog No. 19910610 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-610 2X-Large
19-910-608 Large
19-910-609 X-Large
19-910-611 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-910-610 Supplier Medicom™ Supplier No. 40.036C/05
This item is not returnable. View return policy
Medicom™ Hopen™ HOP'CLEAN Hygiene Coveralls in SMS-Polypropylene limits particulate release (lint-free)
  • HOP'CLEAN Hygiene Coveralls protect the user from activity related soiling
  • Pleasant and light, SMS Polypropylene material is breathable but offers a better seal than a simple polypropylene
  • Suit offers good resistance to tearing and abrasion
  • Permeable to air and water vapors
Specifications
Product Type Single-use Coveralls
Size 2X-Large
Gender Unisex
Material Polypropylene, SMS
Color Blue
Disposable Single-use
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Collared
Seam Type Stitched
Sterility Non-sterile
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
No. of Pockets None