Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Single Use Hygiene Gown

Catalog No. 19910642 Shop All Medicom Products
Change view
Click to view available options
Closure Type:
Press Studs
Zipper
Size:
Medium
Large
X-Large
2X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type Size
19-910-642 Zipper X-Large
19-910-634 Press Studs Medium
19-910-635 Press Studs Large
19-910-636 Press Studs X-Large
19-910-637 Press Studs 2X-Large
19-910-641 Zipper Large
19-910-643 Zipper 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-910-642 Supplier Medicom™ Supplier No. 40.630/04
This item is not returnable. View return policy

Medicom™ Hopen™ Single Use Hygiene Gown without pockets, is microporous and waterproof

  • Reduces contamination of the operator, but also sensitive products and processes in a protected atmosphere
  • Protects against radioactive particles and infectious agents
  • Microporous, Anti-Static Hygiene Gown is comfortable and light
  • Polypropylene gown lined with laminated polyethylene for a waterproof, hydrophobic seal
  • Available in mulitple sizes, and closure types

Specifications

Gender Unisex
Packaging Type Individual, Bag
No. of Pockets None
Size X-Large
For Use With (Application) Category 1 Personal Protective Equipment
Product Type General Purpose Gown
Certifications/Compliance (Europe) EN 1149-5
Construction Made in China
Material Polypropylene
Color White
Closure Type Zipper
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Finish Microporous, Waterproof
Seam Type Sewn
Resistance Radioactive Particles, Infectious Agents
Certifications/Compliance ISO 9001
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.