Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Pocketed, Single Use Hygiene Gown

Catalog No. 19910640 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
White
Closure Type:
Press Studs
Zipper
Size:
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type Size
19-910-640 Blue Zipper 2X-Large
19-910-638 Blue Zipper Large
19-910-639 Blue Zipper X-Large
19-910-631 White Press Studs Large
19-910-632 White Press Studs X-Large
19-910-633 White Press Studs 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-910-640 Supplier Medicom™ Supplier No. 40.601/05
This item is not returnable. View return policy

Medicom™ Hopen™ Pocketed, Single Use Hygiene Gown

  • Single use hygiene gowns protect the working environment from particles coming from the body of the operator (dander, hair, sweating ...)
  • They are suitable to HACCP procedures.
  • They allow the operator to protect himself from the risks of soiling related to the activity he carries out.

Specifications

Certifications/Compliance (Europe) EN 1149-5
Size 2X-Large
Packaging Type Individual, Bag
No. of Pockets 2
Gender Unisex
For Use With (Application) Category 1 Personal Protective Equipment
Product Type General Purpose Gown
Construction Made in China
Material Polypropylene
Color Blue
Closure Type Zipper
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Finish Microporous, Anti-Static
Seam Type Sewn
Resistance Waterproof
Certifications/Compliance ISO 9001
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.