Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Pocketless Hygiene Gown

Catalog No. 19910564 Shop All Medicom Products
Change view
Click to view available options
Color:
Green
Red
White
Size:
Medium
Large
X-Large
2X-Large
3X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-910-564 Red 2X-Large
19-910-566 Green X-Large
19-910-567 Green 2X-Large
19-910-563 Red X-Large
19-910-565 Red 3X-Large
19-910-558 White Medium
19-910-559 White Large
19-910-560 White X-Large
19-910-561 White 2X-Large
19-910-562 White 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-910-564 Supplier Medicom™ Supplier No. 40.301R/05
This item is not returnable. View return policy

Medicom™ Hopen™ Pocketless Hygiene Gown available in three distinct colors

  • The Pocketless Hygiene gown protects the working environment from particles coming from the body of the operator
  • Allows the user to be protected from the risks of soiling related to the activity being carried out
  • Pleasant and light, polypropylene fabric is extremely breathable
  • Collared model without flap and elasticated at the wrists

Specifications

No. of Pockets None
Gender Unisex
For Use With (Application) Category 1 Personal Protective Equipment
Packaging Type Individual, Bag
Size 2X-Large
Construction Made in China
Product Type Hygiene Gown
Material Polypropylene
Color Red
Closure Type Press Studs
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Seam Type Sewn
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.