Learn More
Medicom™ Hopen™ Safe+Mask™ Premier Mask
Catalog No. 19910542 Shop All Medicom Products
Change view
Click to view available options
White
Catalog No.
19-910-542 White
1 options
This item is not returnable. View return policy
Catalog No. 19-910-542 Supplier Medicom™ Supplier No. DB3050
This item is not returnable. View return policy
Medicom™ Hopen™ Safe+Mask™ Premier Mask is splash resistant, and reduces fog formation
  • Meets EN 14683, Type IIR Standards
  • Recommended for use in clean room environments
  • Adjustable nosepiece with synthetic elastics
  • Ensures a good fit for a wide variety of morphologies
  • Splash resistant
Specifications
Product Type General Purpose Face Mask
Color White
Material Polypropylene
Sterility Non-sterile
Disposable Single-use