Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Single Elastic Bouffant Cap

Catalog No. 19910696 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-910-696 Blue
19-910-695 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-910-696 Supplier Medicom™ Supplier No. 70.102

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Medicom™ Hopen™ Single Elastic Bouffant Cap provides full-head coverage to ensure user retention

  • Simple elastic model provides the user with good support by covering the entirety of their hair
  • Possibility of identification by color code
  • Intended to be worn by patients and caregivers in the operating room
  • Can be used as an object for controlling the environment in HACCP and various other industries

Specifications

Material Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
For Use With (Equipment) Operating Room PPE
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Packaging Type Bag
Dimensions (L x W) 44 x 50 cm
Construction Elasticated with Welded Ends
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.