Description
- Filtering half mask, efficency class FFP2 (D)
- Protects users against moderate levels of fine dust particles, and oil or water based mist
- Hazards are initially identified and evaluated by the employer as stipulated in N95 standards
- Typical applications include plastering, cement, sanding, and wood dust
- Prevents against risky infectious diseases, according to national recommendations
- Typical use time: 8 hours
Specifications
Specifications
| Product Type | General Purpose Face Mask |
| Color | Blue |
| Fastening Type | Ties |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Certifications/Compliance (Europe) | EN 14683 |
| Disposable | Single-use |
| Model | M72010S-BL |