Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Visitor Gown

Catalog No. 19910581 Shop All Medicom Products
Change view
Click to view available options
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-910-581 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-910-581 Supplier Medicom™ Supplier No. 50.001
This item is not returnable. View return policy

Medicom™ Hopen™ Visitor Gown Category I PPE, made especially for visitors

  • Offers adequate protection that is similar to standard aprons
  • Single use, polyethylene gown is economical
  • Available in one size

Specifications

No. of Pockets None
Product Type Visitor Gown
For Use With (Application) Category I Personal Protective Equipment
Gender Unisex
Color White
Closure Type Snaps
Disposable Single-use
Sterility Non-sterile
Sleeve Style Long Sleeve
Includes Hood
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.