Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Zipper-Closure Hygiene Gown

Catalog No. 19910644 Shop All Medicom Products
Change view
Click to view available options
Size:
X-Large
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-644 X-Large
19-910-645 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-910-644 Supplier Medicom™ Supplier No. 40.635/04
This item is not returnable. View return policy

Medicom™ Hopen™ Zipper-Closure Hygiene Gown with no pockets, suitable for multiple uses

  • This Zipper-Closure Hygiene Gown protects the operator from work-related contamination
  • Also protects against sensitive products and processes that could be contaminated in a protected atmosphere
  • Pleasant and light, polypropylene fabric is breathable
  • Individually packaged for controlled hygiene and distribution
  • Without pockets to prevent falling objects

Specifications

Construction Made in China
Packaging Type Individual, Bag
No. of Pockets None
Size X-Large
For Use With (Application) Category 1 Personal Protective Equipment
Gender Unisex
Product Type General Purpose Gown
Material Polypropylene
Color Blue
Closure Type Zipper
Disposable Reusable
Sterility Non-sterile
Cuff Style Elastic
Seam Type Sewn
Resistance Radioactive Particles, Infectious Agents
Certifications/Compliance ISO 9001
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.