Learn More
Stoelting™ Hopkins Bulldog Clamp
SDP
Catalog No. 10001379 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Slight curve
Straight
Length (English):
1.5 in.
1.85 in.
2 in.
2.5 in.
3 in.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Length (English)
10-001-379 Straight 2.5 in.
10-000-481 Straight 1.5 in.
10-000-482 Slight curve 1.5 in.
10-000-483 Straight 1.85 in.
10-001-378 Curved 2 in.
10-001-380 Curved 2.5 in.
10-001-381 Straight 3 in.
10-001-382 Curved 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 10-001-379 Supplier Stoelting™ Supplier No. 5212045
Add to Cart
Add to Cart
Stoelting™ Hopkins Bulldog Clamp is useful in surgeries that require precise blood stoppage
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.
Specifications
For Use With (Application) Neuroscience
Length (English) 2.5 in.
Model Straight