Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOR5'Beta13 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5100938
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100938 Supplier Thermo Scientific Supplier No. PA5100938

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HOR5'Beta13.

HOR5'Beta13 gene ontology (GO) annotation includes G protein-coupled receptor activity, olfactory receptor activity and sensory reception of smell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOR5'Beta13
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Host Species Rabbit
Immunogen A synthesized peptide derived from human HOR5'Beta13, corresponding to a region within C-terminal amino acids.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.