Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Pentra™ 400 Clinical Chemistry Analyzer Accessories: Reagent Cups
GSA_VA

Catalog No. 23600504 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Capacity (Metric):
4 mL
10 mL
15 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
23-600-504 10 mL
23-600-506 15 mL
23-600-505 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-600-504 Supplier HORIBA Supplier No. 1221034626
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with Pentra 400 Clinical Chemistry Analyzer

  • Available in 4, 10 and 15mL sizes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type Reagent Cup
Capacity (Metric) 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.