Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Pentra™ 400 Clinical Chemistry Analyzer Accessories: Sample Cups
GSA_VA

Catalog No. 23600496 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Color:
Blue
Green
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-600-496 White
23-600-494 Blue
23-600-495 Green
23-600-497 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-600-496 Supplier HORIBA Supplier No. 1220001767
Add to Cart
Add to Cart

For use with Pentra 400 Clinical Chemistry Analyzer

  • Available in multiple colors

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type Sample Cup
For Use With (Equipment) ABX Pentra™ 400
Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.