Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Reagents for ABX Pentra C400

Catalog No. 23600700 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Type:
CO2 Calibrator
CO2 Control
General Chemistry Reagent
Reference Reagent
Form:
Liquid
Ready-to-Use (RTU)
Quantity:
125 Tests
280 mL
3 x 3mL
4 x 3mL + 4 x 3mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Form Quantity
23-600-700 General Chemistry Reagent Liquid 125 Tests
23-601-500 CO2 Calibrator Ready-to-Use (RTU) 3 x 3mL
23-601-501 CO2 Control Ready-to-Use (RTU) 4 x 3mL + 4 x 3mL
23-601-000 Reference Reagent Ready-to-Use (RTU) 280 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-600-700 Supplier HORIBA Supplier No. 1220001871
Add to Cart
Add to Cart
This item is not returnable. View return policy

FDA cleared Liquid Chemistry

  • With extended linearity and increased stability
  • Standardized specific proteins
  • Purified and concentrated antiserums

Compliance

Complies with international recommendations (IFCC)

Specifications

Type General Chemistry Reagent
For Use With (Application) Serum, Plasma, Whole Blood or Hemolysate (HbA1c), and Urine (Microalbumin)
Quantity 125 Tests
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.