Learn More
HORIBA Enzyme Assays for ABX Pentra™ 400 Clinical Chemistry Analyzers
Shop All ABX Diagnostics, Inc. Products
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Type | Lipase CP Reagent |
| Formulation | Colorimetric |
| Includes | 24mL R1 includes good's Buffer, Taurodesoxycholate, Desoxycholate, Calcium Chloride, Colipase, detergent, preservative, 7mL R2 includes Tartrate Buffer, Taurodesoxycholate, color substrate, coemulgator, Stabilizer, preservative |
| Quantity | 100 Tests |