Learn More
HORIBA General Chemistry Reagents for ABX Pentra™ 400 Clinical Chemistry Analyzers
Shop All ABX Diagnostics, Inc. Products
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Type | Phosphorus CP Substrate |
| Formulation | Ammonium Molybdate UV |
| Quantity | 100 Tests |