Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBF Bioscience Horizontal Mini-Gel Productivity Packages
SCI_ED

Catalog No. S22871 Shop All CBS Scientific Products
Change view
Click to view available options
Gel Dimensions:
L x W: 8.8 x 5.7 cm
Volume (Metric) Buffer:
150 mL
Dimensions (L x W):
Gel Tray I.D.: 8.8 x 5.2 cm
Dimensions (L x W x H):
18.5 x 8.5 x 13 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gel Dimensions Volume (Metric) Buffer Dimensions (L x W) Dimensions (L x W x H)
S22871 L x W: 8.8 x 5.7 cm 150 mL Gel Tray I.D.: 8.8 x 5.2 cm 18.5 x 8.5 x 13 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S22871 Supplier MBF Bioscience Supplier No. GCMHU102

missing translation for 'validMainframeMsg'

Allows casting second gel while first is running for increased efficiency.

CBS Mini Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.
  • Platinum electrode cassettes can be easily removed for cleaning or replacement
  • High quality, heavy duty construction with vast array of accessory options
  • Winged gel trays for easy handling. No more sticking fingers into slippery buffer to extract gel tray
  • Addition of external gel caster and additional gel tray allows researcher to use time more efficiently
  • Tapered baffles allow easy in situ gel formation with or without gel tray
  • Run up to 32 samples per gel. Two slots in each gel tray
  • Combs are printed with catalog number and thickness for easy identification
  • Choice of double-sided adjustable height combs gives researcher more options
  • Optional PTFE-coated aluminum combs for low-friction comb removal and accelerated well formation
  • Horizontal Mini-Gel Systems are available in four vivid colors; green, yellow, blue or fluorescent pink
  • Color-coded horizontal systems are recommended to differentiate exclusive use by specific lab or for specific purpose
Applications: Seperation and identification of DNA fragments, RNA fragments, PCR Products and synthetic oligonucleotides

Specifications

Certifications/Compliance CE
Color Standard Clear
Description Compatible Gel Tray: MGT-122
Type Mini Gel Electrophoresis System
Gel Dimensions L x W: 8.8 x 5.7 cm
Volume (Metric) Buffer 150 mL
Warranty 3 Years
Dimensions (L x W) Gel Tray I.D.: 8.8 x 5.2 cm
Dimensions (L x W x H) 18.5 x 8.5 x 13 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.