Learn More
Poltex Horizontal Tip Organizer-3 Pocket with Wall Mount
SDP
Catalog No. 21100517
Change view
Click to view available options
Includes:
Both Brackets
None
Pipette Aid Bracket
Pipettor Bracket
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-100-517 None
21-100-518 Pipette Aid Bracket
21-100-519 Both Brackets
21-100-520 Pipettor Bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-100-517 Supplier Poltex Supplier No. HZORG3W
Add to Cart
Add to Cart
Poltex Horizontal Tip Organizer-3 Pocket is a horizontally mounted Pipette Tip Organizer for inside BSC, or other convenient area.
Horizontally mounted Pipette Tip Organizer for inside BSC, or other areas
Super durable and cleanable PETG construction
3 pockets
Optional add-ons:
  • Pipette-Aide bracket
  • 4 Pipettor bracket
  • Both brackets
Screws and anchors included to mount organizer
Specifications
Autoclavable Not Autoclavable
Color Clear
Holds Pipette Tips
Material PETG
Positions 3
Shape Rectangular
Stackable No
Sterility Non-sterile
Includes None
Product Type Pipette Tip Box Holder
Array 3 x 1