Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hormones: 3,3',5-Triiodo-L-Thyronine

Catalog No. ICN19458583 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1000 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19458583 250 mg
ICN19458580 100 mg
ICN19458590 500 mg
ICN19458591 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19458583 Supplier MP Biomedicals™ Supplier No. 0219458583
Add to Cart
Add to Cart

For research use only. Not intended for therapeutic use.

TRUSTED_SUSTAINABILITY

Specifications

Form Crystalline powder
Product Type Progesterone
Sterility Sterile
pH 9
Quantity 250 mg
Source Human
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.