Learn More
Biomatik Corporation Horse Adrenomedullin (ADM) ELISA Kit
Supplier:  Biomatik Corporation EKU08521
Encompass_Preferred
Horse Adrenomedullin (ADM) ELISA Kit for detecting ADM in serum, plasma, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 12.35-1000pg/ml. Sensitivity: 4.92pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9N0F0. Target Synonyms: AM; PAMP; Proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5419