Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Horse anti-Mouse IgG ImmPRESS(TM) Secondary Antibody [HRP Polymer], R&D Systems™
SDP

Catalog No. MP7500NB Shop All Novus Products
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP7500NB 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP7500NB Supplier R&D Systems Supplier No. MP7500NB15ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.Horse Polyclonal Antibody

IgG ImmPRESS(TM) Polyclonal antibody specifically detects IgG ImmPRESS(TM) in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, In-situ Hybridization

Specifications

Antigen IgG ImmPRESS(TM)
Applications ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, In Situ Hybridization (ISH)
Classification Polyclonal
Conjugate HRP
Host Species Equine
Immunogen Mouse/Rabbit IgG
Purification Method Affinity purified
Quantity 15 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.