Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Horse IgG, Fc Native Protein
SDP

Catalog No. NBP197034 Shop All R&D Systems Products
Change view
Click to view available options
2mg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP197034 2mg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197034 Supplier Novus Biologicals™ Supplier No. NBP197034
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Native Horse IgG, Fc. Source: Serum The Horse IgG, Fc Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page.

Specifications

For Use With (Application) ELISA, Immunohistochemistry, SDS-PAGE, Western Blot
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Name Horse IgG, Fc Protein
Storage Requirements Store at 4C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Equine
Purity or Quality Grade Multi-step
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.