Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Horse Serum Albumin Native Protein
SDP

Catalog No. NBP250615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250615 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250615 Supplier Novus Biologicals™ Supplier No. NBP250615
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Native Horse Serum Albumin. Source: Serum The Horse Serum Albumin Native Protein has been validated for the following applications: Western Blot, ELISA.

Specifications

Accession Number P35747
For Use With (Application) ELISA, Western Blot
Formulation 0.01M Sodium Phosphate, 0.14M Sodium Chloride, pH 7.4
Gene ID (Entrez) 100034206
Quantity 10 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias ALB Equ c 3, Serum Albumin
Common Name Horse Serum Albumin
Conjugate Unconjugated
Form Lyophilized
Purity or Quality Grade Multi-step
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.