Learn More
Cytiva Spare Parts and Accessories for 142 and 293 mm Disc Filter Holder -
Supplier:  Cytiva 15206
Encompass_Preferred
Convenient Sizes and Durable Materials for Filtration of All Your Laboratory Samples-Design optimizes use of filter area with 15% more EFA than most competitive units, giving higher flow rates and extending the filter's service life.-Sanitary connections are easy to use and suitable for pharmaceutical applications.-Corrosion-resistant type 316 stainless steel construction provides broad chemical resistance. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3210
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.